Order Entry
United States
ContactUsLinkComponent
Mammalian 293-ES Cells
Mammalian 293-ES Cells
Catalog # 77705-764
New Product
CAS Number:  
Mammalian 293-ES Cells
Catalog # 77705-764
Supplier Number:  94-007F
New Product
CAS Number:  

Specifications

  • Cat. No.:
    77705-764

Specifications

About this item

Expression Systems offers 293 and CHO mammalian cell lines adapted to serum-free suspension culture in ESF SFM.

  • Capable of producing high levels of AAV vectors
  • Capable of high levels of recombinant protein expression
  • Good growth in adherent or suspension culture
  • Quality and performance testing
  • Organism: Homo sapiens, human
  • Tissue: Embryonic Kidney
  • Cell Type: Epithelial

Biosafety Level 2: Cells contain adenovirus. Biosafety classification is based on U.S. Public Health Service Guidelines, it is the responsibility of the customer to ensure that their facilities comply with biosafety regulations for their own country.

293-ES cells are derived from the human embryonic kidney HEK 293 cell line. 293-ES cells are adapted for suspension culture in ESF SFM and are available as a frozen vial or suspension culture.

The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.

Culture suspension shipments immediately, liquid nitrogen (vapor phase) for frozen vials.

Delivery: Priority overnight.