Order Entry
United States
ContactUsLinkComponent
S2 Drosophila Cells
S2 Drosophila Cells
Catalog # 77705-758
New Product
CAS Number:  
S2 Drosophila Cells
Catalog # 77705-758
Supplier Number:  94-005F
New Product
CAS Number:  

Specifications

  • Environmentally Preferable:
  • Cat. No.:
    77705-758

Specifications

About this item

Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF.

  • Recombinant protein expression from transient expression or stable cell line
  • Good growth in adherent or suspension culture
  • Quality and performance testing

The S2 insect cell line is derived from a primary culture of late stage (20 to 24 hours old) Drosophila melanogaster embryos. S2 cells are adapted for suspension culture in ESF 921 or ESF AF and are available as a frozen vial or suspension culture.

The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.

Culture suspension shipments immediately, liquid nitrogen (vapor phase) for frozen vials.

Delivery: Priority overnight.