Standard Twist Knot Wire Wheel, Weiler
Supplier: Safety & Industrial Supplies
Certificates
About this item
Rugged knot-type construction provides aggressive, high-impact action for severe applications.
- Heavy-duty brushing action with some flexibility
Use for cleaning, heavy burr removal, roughening surfaces, and surface conditioning applications. Removes rust, paint, oxides, and scale.
Frequently Bought Together