ContactUsLinkComponent
Properties of Toothpaste - Small Group Learning Kit
Properties of Toothpaste - Small Group Learning Kit
  470231-570
undefined
Properties of Toothpaste - Small Group Learning Kit
  470231-570
 :  IS9350-SGL
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Properties of Toothpaste
  • No. of lab groups:
    5
  • Cat. No.:
    470231-570
Create your own Toothpaste.
  • Small group learning kit
  • Student copymasters and teacher guide included
The history of teeth cleaning agents actually goes back thousands of years. Toothpaste as we know it today however is a comparatively recent development. In this activity, students will investigate several properties of toothpaste samples, provided in the kit, including presence of fluoride, pH, abrasiveness, and foaming ability. As a second activity, students will use the included materials to create their own toothpaste.