Orders LinkContactUsLinkComponent
General Purpose Dust Removers, MicroCare
General Purpose Dust Removers, MicroCare
Supplier:   MicroCare
General Purpose Dust Removers, MicroCare
Supplier:  MicroCare
Certificates
General Purpose Dust Removers, MicroCare
General Purpose Dust Removers, MicroCare
4 Options Available Below
Product Details & Documents
General Purpose Dust Remover is a high purity electronics-grade cleaner that delivers a dry blast of inert gas to hard to reach surfaces. Use to remove dust, particles, and moisture from any surface.
  • Cleans dust & grit from keyboards, optics, machinery & circuitry
  • Trigger spray allows easy single-handed operation
  • Plastic-safe, nonflammable and leaves no residue
  • High-pressure
  • Multiple package sizes available
Clean hard-to-reach areas like keyboards or pin connectors. Use to quickly pre-clean surfaces by removing layers of loose grime and dust before using a solvent cleaner. Also, use to remove slow-drying solvents from under surface mount components or on circuit boards, connectors or optics.
Product Family Options
Product Information
  • Volume
    Availability
    Price
  • 76020-202
    General purpose dust remover, StatZap compatible
    General purpose dust remover, StatZap compatible
    284 g (10 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $107.82
    $107.82
    Log in for availability
    Case of 12
    Case of 12 - $1,293.84
    $1,293.84
    Each
       $107.82
    Case of 12
       $1,293.84
    Supplier Number: MCC-DSTZ
  • 76020-200
    General purpose dust remover, jumbo size
    General purpose dust remover, jumbo size
    397 g (14 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $108.95
    $108.95
    Log in for availability
    Case of 12
    Case of 12 - $1,307.40
    $1,307.40
    Each
       $108.95
    Case of 12
       $1,307.40
    Supplier Number: MCC-DST14A
  • 76020-204
    General purpose dust remover, StatZap compatible, jumbo size
    General purpose dust remover, StatZap compatible, jumbo size
    397 g (14 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $65.19
    $65.19
    Log in for availability
    Case of 12
    Case of 12 - $782.28
    $782.28
    Each
       $65.19
    Case of 12
       $782.28
    Supplier Number: MCC-DSTZ14
  • 76020-198
    General purpose dust remover, 360° invertible
    General purpose dust remover, 360° invertible
    227 g (8 oz.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $65.42
    $65.42
    Log in for availability
    Case of 12
    Case of 12 - $785.04
    $785.04
    Each
       $65.42
    Case of 12
       $785.04
    Supplier Number: MCC-DST08A