ContactUsLinkComponent
Friedrichs Condensers, with Removable Hose Connections, Chemglass
  47733-780
 :  Chemglass
Small Business Enterprise
undefined
Friedrichs Condensers, with Removable Hose Connections, Chemglass
  47733-780
 :  Chemglass
 :  CG-1210-HC-01
Small Business Enterprise
  • [ST] joint:
    24/40
  • Description:
    Friedrichs Condenser, Removable Hose Connection, GL-14 Screw Caps
  • Cat. No.:
    47733-780
  • Overall height:
    330 mm
  • Supplier no.:
    CG-1210-HC-01
This Friedrichs condenser is constructed with detachable hose connections.
The connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Hose connections have an O.D. of 8 mm and an I.D. of 4 mm.
 : Supplied complete with two GL-14 open top screw caps. See CG-198 and CG-195 for replacement GL fittings.