Proto® Super Heavy-Duty Offset Slugging Wrenches, Essendant
Supplier: Safety & Industrial Supplies
Certificates
About this item
Heavy box walls provide greater torque transmission
- Black oxide finish
- Box opening
- Forged alloy steel
- Inch measuring system
- Off-set handle
High strength rectangular design withstands heavy sledging. Straight handle resists springing for easier nut turning. For close quarter work where hammer or sledge is required to loosen frozen nuts or set large fasteners.
Box opening remains on fastener when wrench is struck.
Frequently Bought Together