Proto® Torqueplus™ Flex Impact Sockets ¹/₂", ORS Nasco
76190-006
- Cat. No.:76190-006
- Overall length:2²¹⁄₃₂"
- Head width:47/64" (opening side); 1 9/64" (drive side)
- Opening size:½"
- Drive size:1/2"
- No. of points:6
Reduces risk of fastener rounding.
- Box tip
- Pin locking
- Square drive
- Black oxide finish
- Forged alloy steel
- Inch measuring
Chamfered service ends. Patented "Pinless" design. Thru hole and groove accept retaining ring and pin, when securing to power tool. Applies more torque to fastener. 65° turning angle. Optimized socket wall thickness. Concentric openings.