ContactUsLinkComponent
Proto® Drive Impact Universal Joint Sockets ¹/₂", ORS Nasco
  76189-812
Proto® Drive Impact Universal Joint Sockets ¹/₂", ORS Nasco
  76189-812
 :  577-74470P
  • Height:
    1.4"
  • Cat. No.:
    76189-812
  • Width:
    1.4"
  • Drive size:
    1/2"
Tight tolerances for snug fit on fastener.
  • Black oxide finish
Used wherever powered drive tools are used to install or remove fasteners. Designed to withstand the high torque and force associated with impact tools in repetitive application. Features two through holes and a groove to accept a Proto Retaining Ring or pin and O-ring to help secure the socket to the drive tool anvil. Position at appropriate angle to turn fasteners where obstructions prevent a straight application. Ball and spring design which allows for a 30° working angle. Smooth 360° rotation.