ContactUsLinkComponent
Proto® Torqueplus™ Impact Sockets, ¹/₂", ORS Nasco
Catalog # 76189-814
Proto® Torqueplus™ Impact Sockets, ¹/₂", ORS Nasco
Catalog # 76189-814
Supplier Number:  577-7450H
Specifications
  • Cat. No.:
    76189-814
  • Overall length:
    2¼"
  • Head width:
    2 1/32" (opening side); 1 5/8" (drive side)
  • Opening size:
    1⁹⁄₁₆"
  • Drive size:
    1/2"
  • No. of points:
    6
Specifications
About this item
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded or rusted fasteners
  • Forged alloy steel
  • Square drive
  • Black oxide finish
  • Pin locking
  • Inch measuring
Proto Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.