Proto® TorquePlus™ Thin Wall Deep Impact Sockets, Stanley® Products
Supplier: Safety & Industrial Supplies
Certificates
About this item
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
- Forged alloy steel
- Square drive
- Black oxide finish
Proto Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.
Frequently Bought Together