Proto® ProtoGrip™ High Strength Magnetic Impact Sockets, ORS Nasco
Allows room for more magnet material, helping increase total magnetic strength.
- Forged alloy steel
- Black oxide finish
- Square drive
- Pin locking
Designed to easily pick up heavy fasteners and hold them tightly during use. Impact sockets with high impact strength are used in automated and hand impact socket application. ProtoGrip™ Magnetic Sockets require no bushings to hold the magnet in place. ProtoGrip™ Magnetic Sockets use high strength magnets made from a rare earth magnet alloy/nylon blend. Strong magnetic hold helps increase efficiency when installing fasteners in awkward or hard-to-reach locations.
Frequently Bought Together