Proto® 15 Piece Crowfoot Wrench Sets, ORS Nasco
Supplier: Safety & Industrial Supplies
Certificates
About this item
Anti-Slip Design open end (ASD) has patented locking grooves that help prevent the wrench from slipping off the fastener when turning.
- Open opening type
- Forged alloy steel
- Inch measuring system
- Full polish
Crowfoot wrenches are used in hard to reach, restricted access situations where a standard wrench or ratchet cannot reach. Full polish finish provides corrosion resistance and easy-to-clean surfaces. Locking hole feature helps keep the crowfoot wrench engaged on the extension during use, reducing downtime due to detachment of crowfoot in awkward spaces.
Frequently Bought Together