ContactUsLinkComponent
Proto® Crowfoot Wrench Sets, 27 Piece, ORS Nasco
Proto® Crowfoot Wrench Sets, 27 Piece, ORS Nasco
Catalog # 76190-708
Proto® Crowfoot Wrench Sets, 27 Piece, ORS Nasco
Catalog # 76190-708
Supplier Number:  577-49002CF
Specifications
  • Cat. No.:
    76190-708
  • Opening size:
    ⅜"; ⁷⁄₁₆"; ½"; ⁹⁄₁₆"; ⅝"; ¹¹⁄₁₆"; ¾"; 1³⁄₁₆; ⅞"; ¹⁵⁄₁₆"; 1"; 1¹⁄₁₆"; 1⅛"; 1³⁄₁₆"; 1¼"; 1⁵⁄₁₆"; 1⅜"; 1⁷⁄₁₆"; 1½"; 1⁹⁄₁₆"; 1⅝"; 1¹¹⁄₁₆"; 1¾"; 1¹³⁄₁₆"; 1⅞"; 1¹⁵⁄₁₆"; 2"
  • Drive size:
    3/8"
Specifications
About this item
Anti-Slip Design open end (ASD) has patented locking grooves that help prevent the wrench from slipping off the fastener when turning.
  • Locking hole on 3/8" drive crowfoot wrenches available on fractional openings 1" and larger
  • Typically used in lower torque applications
  • Full polish finish
  • Open opening type
  • Inch measuring systemx
  • Forged alloy steel
Crowfoot wrenches are used in hard to reach, restricted access situations where a standard wrench or ratchet cannot reach. Full polish finish provides corrosion resistance and easy-to-clean surfaces. Locking hole feature helps keep the crowfoot wrench engaged on the extension during use, reducing downtime due to detachment of crowfoot in awkward spaces. Locking hole on 3/8" drive crowfoot wrenches available on fractional openings 1" and larger. Typically used in lower torque applications.