Proto® TorquePlus™ Sockets, Stanley® Products
Catalog # 76190-614
Supplier: Safety & Industrial Supplies
Specifications
- Description:Sockets
- Cat. No.:76190-614
- Overall length:5⅝"
- Head width:5" (opening side); 5" (drive side)
- Opening size:2⅞"
- Drive size:2 1/2"
- No. of points:6
Specifications
About this item
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
- inch measuring system
- Forged alloy steel
- Pin locking type
- Square drive
- Black oxide finish
Proto Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.