ContactUsLinkComponent
Proto® Torqueplus™ 12-Point Combination Wrenches, Satin Finish, ORS Nasco
Proto® Torqueplus™ 12-Point Combination Wrenches, Satin Finish, ORS Nasco
Catalog # 76190-162
Proto® Torqueplus™ 12-Point Combination Wrenches, Satin Finish, ORS Nasco
Catalog # 76190-162
Supplier Number:  577-1211A
Specifications
  • Description:
    Combination Wrenches
  • Cat. No.:
    76190-162
  • Overall length:
    5⅝"
  • Offset:
    15° box side
  • Head width:
    25/32" (open); 17/32" (box)
  • Head thickness:
    ⁷⁄₃₂" (box), ³⁄₁₆" (open)
  • Opening size:
    ¹¹⁄₃₂"
  • No. of points:
    12
  • Head angle:
    15° open side
Specifications
About this item
Torqueplus™ fastening system drives off fastener body, more easily removing damaged fasteners.
  • Oval handle
  • Open/box opening type
  • Inch measuring system
  • Forged alloy steel
  • Satin finish
Reduces risk of fastener rounding. Applies more torque to fastener. Unique arc design increases surface contact. Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury. Shank is positioned at the center of box end, reduces the tendency of the wrench to cock.