Venturi V07-General Purpose Bench Fume Hood with Horizontal Sash, KMER Liner, Kewaunee Scientific
Supplier: Avantor
Certificates
About this item
The Supreme Air Venturi provides the safest user friendly fume hood manufactured today.
- Unparalled Venturi enhanced containment technology
- High performance/Energy efficient
- Unique ultra-smooth sash system
- Exceptional viewing height
- Flush airfoil
- LED lights with color and intensity adjustability
- Electromechanical sash stop
The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stop, ultra-smooth sash system, and push button sash management.
Certifications: UL and cUL listed. Tested to ASHRAE 110, NIH03-112C, ASTM D552, and NFPA 45.
Ordering information: Work top and base cabinets not included with fume hood.
Frequently Bought Together