Order Entry
United States
ContactUsLinkComponent
 
Mouse Recombinant Stem cell factor
 
Mouse Recombinant Stem cell factor

 

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Mouse
  • Endotoxin Content:
    ≤1 EU/μg protein measured by kinetic LAL analysis.
  • Biological Activity:
    The activity is determined by the dose-dependent proliferation of human TF-1 cells which is typically observed at concentrations <10 ng/ml.
  • Protein/Peptide Name:
    Stem cell factor
  • Purity:
    >98% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
  • Molecular Weight:
    ~18.3 kDa
  • Formulation:
    Lyophilized from a concentrated (1mg/ml) solution in water containing 10 mM Na2PO4, pH 6.5 Sterile filtered.

 

Frequently Bought Together