Dri-Shield® Moisture Barrier Bag, 2000 Series, SCS, Desco Industries
Catalog # 89024-310
Supplier: Desco
Specifications
- Dimensions:3×5"
- Thickness:3.6 mil.
- Cat. No.:89024-310
- Packaging:100 bags/pack
- Supplier no.:70035
Specifications
About this item
This aluminized moisture barrier bag is designed to provide packaging for ESD and moisture sensitive items, both inside and outside an ESD protected area.
- Heat sealable and suitable for vacuum packaging
- Printed with ESD and moisture warning symbols and a lot code for traceability
- Manufactured from a laminate of multiple layers of aluminized polyester and polyethylene
Metal layers are intended to provide shielding of Electrostatic discharge (ESD) and to help minimize the penetration of electric field while providing moisture protection. Dissipative inner and outer surfaces allows bag to be grounded when placed on a grounded surface or handled by a grounded operator.
Certifications: Meets ANSI/ESD S20.20, ANSI/ESD S541, EIA 541, JESD625-A, and EIA 583.