Temp-Chex Laboratory Thermometers, Streck Laboratories
Supplier: Streck Laboratories
Certificates
About this item
Thermometers are ideal for monitoring refrigerators, freezers, incubators and room temperatures.
- Mercury free
- Scaled to 1/2° increments (Ultra-Low thermometer features 1° gradation)
- Available in Enviro-Safe® and red spirit
Thermometers are contained in a vial of propylene glycol to buffer temperature variations. Each thermometer is enclosed in a plastic case to protect the thermometer and contain the contents if broken. Attach to refrigerator, freezer or incubator walls magnetically or adhesively. Enviro-Safe® thermometers utilize only biodegradable, nontoxic and nonflammable components.
Certifications: Each thermometer is engraved with a serial number traceable to National Institute of Standards and Technology (NIST) standards.
Delivery: A wall chart for daily temperature recording is included with each Temp-Chex.
Frequently Bought Together