Anti-Troponin T type 1 (slow skeletal) Mouse Polyclonal Antibody
Catalog # 103333-316
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Troponin T type 1 (slow skeletal)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:7138
- Antigen Synonyms:MGC104241|slow skeletal muscle|Slow skeletal muscle troponin T|troponin T type 1 (skeletal|ANMtroponin T|TNTS|troponin-T1|slow|skeletal|slow)|TNTtroponin T1|FLJ98147|STNT
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TNNT1 (NP_003274, 1 a.a. - 251 a.a.) full-length human protein. MSDTEEQEYEEEQPEEEAAEEEEEEEERPKPSRPVVPPLIPPKIPEGERVDFDDIHRKRMEKDLLELQTLIDVHFEQRKKEEEELVALKERIERRRSERAEQQRFRTEKERERQAKLAEEKMRKEEEEAKKRAEDDAKKKKVLSNMGAHFGGYLVKAEQKRGKRQTGREMKVRILSERKKPLDIDYMGEEQLREKAQELSDWIHQLESEKFDLMAKLKQQKYEINVLYNRISHAQKFRKGAGKGRVGGRWK
- Purification:Protein A purified
- Cat. No.:103333-316
- Supplier no.:H00007138-B01P
Specifications
About this item
The Troponin T type 1 (slow skeletal) Antibody from Novus Biologicals is a mouse polyclonal antibody to Troponin T type 1 (slow skeletal). This antibody reacts with human. The Troponin T type 1 (slow skeletal) Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Troponin T type 1 (slow skeletal)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human