Anti-Thioredoxin Mouse Monoclonal Antibody [clone: 2A7]
103333-544
- Antibody Type:Primary
- Antigen Name:Thioredoxin
- Clonality:Monoclonal
- Clone:2A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:7295
- Antigen Synonyms:TRDX|Trx|Surface-associated sulphydryl protein|MGC61975|thioredoxin|TRXDKFZp686B1993|ATL-derived factor|TRX1|SASP|ADF
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TXN (AAH03377, 1 a.a. - 105 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVKQIESKTAFQEALDAAGDKLVVVDFSATWCGPCKMIKPFFHSLSEKYSNVIFLEVDVDDCQDVASECEVKCMPTFQFFKKGQKVGEFSGANKEKLEATINELV
- Purification:IgG purified
- Cat. No.:103333-544
- Supplier no.:H00007295-M01
The Thioredoxin Antibody (2A7) from Novus Biologicals is a mouse monoclonal antibody to Thioredoxin. This antibody reacts with human. The Thioredoxin Antibody (2A7) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.
Type: Primary
Antigen: Thioredoxin
Clonality: Monoclonal
Clone: 2A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human