To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Syntaxin Binding Protein 4
- Clonality:Monoclonal
- Clone:2B12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:252983
- Antigen Synonyms:SynipMGC149829|STX4-interacting protein|MGC50337|syntaxin-binding protein 4|FLJ16496|syntaxin binding protein 4|Syntaxin 4-interacting protein|syntaxin 4 interacting protein
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:STXBP4 (AAH41485, 1 a.a. ~ 246 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.MIGVSFEEAKSIITRAKLSTRLESAWEIAFIRQKSDNIQPENLSCTSLIEASGEYGPQASTLSLFSSPPEILIPKTSSTPKTNNDILSSCEIKTGYNKTVQIPITSENSTVGLSNTDVASAWTENYGLQEKISLNPSVRFKAEKLEMALNYLGIQPTKEQHQALRQQVQADSKGTVSFGDFVQVARNLFCLQLDEVNVGAHEISNILDSQLLPCDSSEADEMERLKCERDDALKEVNTLKWPGQN
- Purification:Protein A purified
- Cat. No.:103347-918
- Supplier no.:H00252983-M03
Specifications
About this item
The Syntaxin Binding Protein 4 Antibody (2B12) from Novus Biologicals is a mouse monoclonal antibody to Syntaxin Binding Protein 4. This antibody reacts with human. The Syntaxin Binding Protein 4 Antibody (2B12) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Syntaxin Binding Protein 4
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human