Anti-Rev-erb A alpha/NR1D1 Mouse Monoclonal Antibody [clone: 5H6]
Catalog # 103337-244
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Rev-erb A alpha/NR1D1
- Clonality:Monoclonal
- Clone:5H6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:9572
- Antigen Synonyms:EAR1nuclear receptor subfamily 1 group D member 1|Rev-erbA-alpha|THRALrev-erbA-alpha|group D|EAR-1|nuclear receptor subfamily 1|V-erbA-related protein 1|member 1|Rev-ErbAalpha|THRA1|HREV
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:NR1D1 (AAH56148, 1 a.a. ~ 615 a.a) full length recombinant protein with GST tag.MTTLDSNNNTGGVITYIGSSGSSPSRTSPESLYSDNSNGSFQSLTQGCPTYFPPSPTGSLTQDPARSFGSIPPSLSDDGSPSSSSSSSSSSSSFYNGSPPGSLQVAMEDSSRVSPSKSTSNITKLNGMVLLCKVCGDVASGFHYGVHACEGCKGFFRRSIQQNIQYKRCLKNENCSIVRINRNRCQQCRFKKCLSVGMSRDAVRFGRIPKREKQRMLAEMQSAMNLANNQLSSQCPLETSPTQHPTPGPMGPSPP
- Purification:IgG purified
- Cat. No.:103337-244
- Supplier no.:H00009572-M05
Specifications
About this item
The Rev-erb A alpha / NR1D1 Antibody (5H6) from Novus Biologicals is a mouse monoclonal antibody to Rev-erb A alpha / NR1D1. This antibody reacts with human. The Rev-erb A alpha / NR1D1 Antibody (5H6) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Rev-erb A alpha/NR1D1
Clonality: Monoclonal
Clone: 5H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human