Anti-PP2C beta/PPM1B Mouse Monoclonal Antibody [clone: 1A3-2A4]
Catalog # 103307-896
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:PP2C beta/PPM1B
- Clonality:Monoclonal
- Clone:1A3-2A4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:5495
- Antigen Synonyms:Protein phosphatase 2C isoform beta|beta isoform|PP2C-beta|MGC21657|protein phosphatase|PP2CBmagnesium-dependent|1B|PP2C-beta-X|EC 3.1.3.16|protein phosphatase 2C-like protein|Mg2+/Mn2+ dependent|protein phosphatase 1B
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PPM1B (AAH12002, 1 a.a. - 192 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSIVLVCFSNAPKVSDEAVKKDSELDKHLESRVEEIMEKSGEEGMPDLAHVMRILSAENIPNLPPGGGLAGKRNVIEAVYSRLNPHRESDGASDEAEESGSQGKLVEALRQMRINHRGNYRQLLEEMLTSYRLAKVEGEESPAEPAATATSSNSDAGNPVTMQESHTESESGLAELDSSNEDAGTKMSGEKI
- Tested Applications:Proximity Ligation Assay, RNA Inhibition
- Purification:IgG purified
- Cat. No.:103307-896
- Supplier no.:H00005495-M01
Specifications
About this item
The PP2C beta / PPM1B Antibody (1A3-2A4) from Novus Biologicals is a mouse monoclonal antibody to PP2C beta / PPM1B. This antibody reacts with human. The PP2C beta / PPM1B Antibody (1A3-2A4) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Proximity Ligation Assay, RNA Inhibition.
Type: Primary
Antigen: PP2C beta/PPM1B
Clonality: Monoclonal
Clone: 1A3-2A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human