Anti-Nitrotyrosine Mouse Monoclonal Antibody [clone: EM-30]
Catalog # 103283-626
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Nitrotyrosine
- Clonality:Monoclonal
- Clone:EM-30
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Antigen Synonyms:Nitrotyrosine
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:NO2-Tyr-CH2-Thyroglobulin
- Purification:Immunogen affinity purified
- Cat. No.:103283-626
- Supplier no.:NBP1-96130
Specifications
About this item
The Nitrotyrosine Antibody (EM-30) from Novus Biologicals is a mouse monoclonal antibody to Nitrotyrosine. This antibody reacts with human. The Nitrotyrosine Antibody (EM-30) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: Nitrotyrosine
Clonality: Monoclonal
Clone: EM-30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human