Anti-ZNF37A Rabbit Polyclonal Antibody
103334-166
- Antibody Type:Primary
- Antigen Symbol:ZNF37A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:7587
- Antigen Synonyms:Zinc finger protein KOX32|zinc finger protein 34|KOX32zinc finger protein 34 (KOX 32)|MGC10520
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ZNF37A (NP_001007095.1, 1 a.a. - 561 a.a.) full-length human protein. MITSQGSVSFRDVTVGFTQEEWQHLDPAQRTLYRDVMLENYSHLVSVGYCIPKPEVILKLEKGEEPWILEEKFPSQSHLELINTSRNYSIMKFNEFNKGGKCFCDEKHEIIHSEEEPSEYNKNGNSFWLNEDLIWHQKIKNWEQSFEYNECGKAFPENSLFLVHKRGYTGQKTCKYTEHGKTCDMSFFITHQQTHPRENHYGNECGENIFEESILLEHQSVYPFSQKLNLTPIQRTHSINNIIEYNECGTFFSEKLVLHLQQRTHTGEKPYECHECGKTFTQKSAHTRHQRTHTGGKPYECHECGKTFYKNSDLIKHQRIHTGERPYGCHECGKSFSEKSTLTQHQRTHTGEKPYECHECGKTFSFKSVLTVHQKTHTGEKPYECYACGKAFLRKSDLIKHQRIHTGEKPYECNECGKSFSEKSTLTKHLRTHTGEKPYECIQCGKFFCYYSGFTEHLRRHTGEKPFGCNECGKTFRQKSALIVHQRTHIRQKPYGCNQCGKSFCVKSKLIAHHRTHTGEKPYECNVCGKSFYVKSKLTVHQRIHLGRNPINVVNEGNYSG
- Purification:Protein A purified
- Cat. No.:103334-166
- Supplier no.:H00007587-D01P
The ZNF37A Antibody from Novus Biologicals is a rabbit polyclonal antibody to ZNF37A. This antibody reacts with human. The ZNF37A Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ZNF37A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human