Anti-ZNF3 Mouse Monoclonal Antibody [clone: 1F7]
Catalog # 103334-114
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:ZNF3
- Clonality:Monoclonal
- Clone:1F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:7551
- Antigen Synonyms:PRDM17|PFM14|PR domain zinc finger protein 17|FLJ12827|zinc finger protein 408
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ZNF3 (NP_060185.1, 1 a.a. - 110 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. METQADLVSQEPQALLDSALLSKVPAFSDKDSLGDEMLAAALLKAKSQELVTFEDVAVYFIRKEWKRLEPAQRDLYRDVMLENYGNVFSLDRETRTENDQEISEDTRSHG
- Purification:IgG purified
- Cat. No.:103334-114
- Supplier no.:H00007551-M08
Specifications
About this item
The ZNF3 Antibody (1F7) from Novus Biologicals is a mouse monoclonal antibody to ZNF3. This antibody reacts with human. The ZNF3 Antibody (1F7) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ZNF3
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human