Anti-Wnt3 Rabbit Polyclonal Antibody
103333-936
- Antibody Type:Primary
- Antigen Symbol:Wnt3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:7473
- Antigen Synonyms:MGC138323|MGC131950|proto-oncogene Wnt-3|WNT-3 proto-oncogene protein|wingless-type MMTV integration site family|member 3|INT4Proto-oncogene Int-4 homolog|MGC138321
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:WNT3 (NP_110380.1, 1 a.a. - 355 a.a.) full-length human protein. MEPHLLGLLLGLLLGGTRVLAGYPIWWSLALGQQYTSLGSQPLLCGSIPGLVPKQLRFCRNYIEIMPSVAEGVKLGIQECQHQFRGRRWNCTTIDDSLAIFGPVLDKATRESAFVHAIASAGVAFAVTRSCAEGTSTICGCDSHHKGPPGEGWKWGGCSEDADFGVLVSREFADARENRPDARSAMNKHNNEAGRTTILDHMHLKCKCHGLSGSCEVKTCWWAQPDFRAIGDFLKDKYDSASEMVVEKHRESRGWVETLRAKYSLFKPPTERDLVYYENSPNFCEPNPETGSFGTRDRTCNVTSHGIDGCDLLCCGRGHNTRTEKRKEKCHCIFHWCCYVSCQECIRIYDVHTCK
- Purification:Protein A purified
- Cat. No.:103333-936
- Supplier no.:H00007473-D01P
The Wnt3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Wnt3. This antibody reacts with human. The Wnt3 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Wnt3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human