Anti-WIF-1 Mouse Polyclonal Antibody
Catalog # 103340-328
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Symbol:WIF-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:11197
- Antigen Synonyms:WIF-1|WNT inhibitory factor 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:WIF1 (AAH18037.1, 1 a.a. ~ 379 a.a) full-length human protein.MARRSAFPAAALWLWSILLCLLALRAEAGPPQEESLYLWIDAHQARVLIGFEEDILIVSEGKMAPFTHDFRKAQQRMPAIPVNIHSMNFTWQAAGQAEYFYEFLSLRSLDKGIMADPTVNVPLLGTVPHKASVVQVGFPCLGKQDGVAAFEVDVIVMNSEGNTILKTPQNAIFFKTCQQAECPGGCRNGGFCNERRICECPDGFHGPHCEKALCTPRCMNGGLCVTPGFCICPPGFYGVNCDKANCSTTCFNGGT
- Purification:Protein A purified
- Cat. No.:103340-328
- Supplier no.:H00011197-B01P
Specifications
About this item
The WIF-1 Antibody from Novus Biologicals is a mouse polyclonal antibody to WIF-1. This antibody reacts with human. The WIF-1 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: WIF-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human