Anti-WBP2NL Mouse Polyclonal Antibody
103347-520
- Antibody Type:Primary
- Antigen Symbol:WBP2NL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:164684
- Antigen Synonyms:postacrosomal sheath WW domain-binding protein|PAWPFLJ26145|WW domain-binding protein 2-like|MGC26816|WBP2 N-terminal like
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:WBP2NL (AAH22546, 1 a.a. ~ 310 a.a) full-length human protein.MAVNQSHTENRRGALIPNGESLLKRSPNVELSFPQRSEGSNVFSGRKTGTLFLTSYRVIFITSCSISDPMLSFMMPFDLMTNLTVEQPVFAANFIKGTIQAAPYGGWEGQATFKLVFRNGGAIEFAQLMVKAASAAARGFPLRTLNDWFSSMGIYVITGEGNMCTPQMPCSVIVYGAPPAGYGAPPPGYGAPPAGYGAQPVGNEGPPVGYRASPVRYGAPPLGYGAPPAGYGAPPLGYGAPPLGYGTPPLGYGAP
- Purification:Protein A purified
- Cat. No.:103347-520
- Supplier no.:H00164684-B01P
The WBP2NL Antibody from Novus Biologicals is a mouse polyclonal antibody to WBP2NL. This antibody reacts with human. The WBP2NL Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: WBP2NL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human