Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Symbol:WASF3/WAVE3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:10810
- Antigen Synonyms:SCAR3WASP family protein member 3|WAVE3Brush-1|wisk|Verprolin homology domain-containing protein 3|KIAA0900WASP family Verprolin-homologous protein 3|WAS protein family|member 3|Protein WAVE-3
- Storage Buffer:PBS (pH 7.0), 1.0% BSA and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human WASF3. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102712-640
- Supplier no.:NBP2-20885
The WASF3 / WAVE3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to WASF3 / WAVE3. This antibody reacts with human. The WASF3 / WAVE3 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: WASF3/WAVE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human