Anti-WARS2 Rabbit Polyclonal Antibody
Catalog # 103338-710
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:WARS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:10352
- Antigen Synonyms:Tryptophan--tRNA ligase|tryptophanyl-tRNA synthetase|EC 6.1.1.2|(Mt)TrpRS|TrpRStryptophan-tRNA ligase|mitochondrial|tryptophan tRNA ligase 2|tryptophanyl tRNA synthetase 2|EC 6.1.1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:WARS2 (NP_957715.1, 1 a.a. ~ 220 a.a) full-length human protein.
- Purification:Protein G purified
- Cat. No.:103338-710
- Supplier no.:H00010352-D01P
Specifications
About this item
The WARS2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to WARS2. This antibody reacts with human. The WARS2 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: WARS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human