Anti-VGLL1 Mouse Polyclonal Antibody
Catalog # 103344-148
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:VGLL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:51442
- Antigen Synonyms:VGL1|TONDU|WUGSC:H_GS188P18.1b|TDUtranscription cofactor vestigial-like protein 1|vestigial like 1 (Drosophila)|Vgl-1|Protein TONDU
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:VGLL1 (NP_057351.1, 1 a.a. ~ 258 a.a) full-length human protein.
- Purification:Protein G purified
- Cat. No.:103344-148
- Supplier no.:H00051442-B02P
Specifications
About this item
The VGLL1 Antibody from Novus Biologicals is a mouse polyclonal antibody to VGLL1. This antibody reacts with human. The VGLL1 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: VGLL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human