To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:VEGFR1/Flt-1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:2321
- Antigen Synonyms:FLT-1|EC 2.7.10|FLT|EC 2.7.10.1|Tyrosine-protein kinase receptor FLT|vascular endothelial growth factor receptor 1|Tyrosine-protein kinase FRT|VEGFR1|fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor)|FRT|VEGFR-1|Vascular permeability factor receptor|Fms-like tyrosine kinase 1
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide made to an internal region of the mouse VEGF Receptor 1 protein (between residues 800-900) [UniProt# P35969]. There is 87% identity between the immunogen used for this production and the VEGF Receptor 2 protein.
- Purification:Immunogen affinity purified
- Cat. No.:103358-310
Specifications
About this item
The VEGFR1 / Flt-1 Antibody [FITC] from Novus Biologicals is a rabbit polyclonal antibody to VEGFR1 / Flt-1. This antibody reacts with human. The VEGFR1 / Flt-1 Antibody [FITC] has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: VEGFR1/Flt-1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human