Anti-UTP14A Mouse Polyclonal Antibody
103339-636
- Antibody Type:Primary
- Antigen Symbol:UTP14A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:10813
- Antigen Synonyms:Antigen NY-CO-16|UTP14|SDCCAG16|NY-CO-16|NYCO16|homolog A (yeast)|Serologically defined colon cancer antigen 16KIAA0266|U3 small nucleolar RNA-associated protein 14 homolog A|FLJ37089|U3 small nucleolar ribonucleoprotein|dJ537K23.3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:UTP14A (NP_006640.2, 1 a.a. ~ 771 a.a) full-length human protein.
- Purification:Protein G purified
- Cat. No.:103339-636
- Supplier no.:H00010813-B01P
The UTP14A Antibody from Novus Biologicals is a mouse polyclonal antibody to UTP14A. This antibody reacts with human. The UTP14A Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: UTP14A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human