Anti-USP52 Mouse Monoclonal Antibody [clone: 6A7]
103337-764
- Antibody Type:Primary
- Antigen Symbol:USP52
- Clonality:Monoclonal
- Clone:6A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:9924
- Antigen Synonyms:PYPAF4|KIAA0710PAB-dependent poly(A)-specific ribonuclease subunit 2|PABP-dependent poly(A) nuclease 2|EC 3.1.13.4|FLJ39360|hPan2|Inactive ubiquitin carboxyl-terminal hydrolase 52|ubiquitin specific protease 52|PAN2 poly(A) specific ribonuclease subunit homolog (S. cerevisiae)|NALP4|ubiquitin specific peptidase 52|USP52|PAN2 polyA specific ribonuclease subunit
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:USP52 (NP_055686, 1099 a.a. ~ 1199 a.a) partial recombinant protein with GST tag.TVYLFHMPRKRMISLRFLAWYFLDLKIQGETHDSIEDARTAL QLYRKYLELSKNGTEPESFHKVLKGLYEKGRKMDWKVPEPEG QTSPKNAAVFSSVLAL
- Purification:IgG purified
- Cat. No.:103337-764
- Supplier no.:H00009924-M01
The USP52 Antibody (6A7) from Novus Biologicals is a mouse monoclonal antibody to USP52. This antibody reacts with human. The USP52 Antibody (6A7) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: USP52
Clonality: Monoclonal
Clone: 6A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human