Anti-TNF RII/TNFRSF1B Rabbit Polyclonal Antibody
103333-304
: Avantor
- Antibody Type:Primary
- Antigen Symbol:TNF RII/TNFRSF1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:7133
- Antigen Synonyms:CD120b|tumor necrosis factor beta receptor|TNFR2TNFR1B|CD120b antigen|p75 TNF receptor|TNFR-II|p75TNFR|TNF-R2|TNF-R-II|TNF-R75|tumor necrosis factor receptor superfamily member 1B|tumor necrosis factor receptor superfamily|p75TBPII|member 1B|Etanercept|soluble TNFR1B variant 1|TNFBRp80 TNF-alpha receptor|TNFR80|Tumor necrosis factor receptor type II|Tumor necrosis factor receptor 2|TNF-RII|tumor necrosis factor binding protein 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TNFRSF1B (NP_001057.1, 1 a.a. - 461 a.a.) full-length human protein. MAPVAVWAALAVGLELWAAAHALPAQVAFTPYAPEPGSTCRLREYYDQTAQMCCSKCSPGQHAKVFCTKTSDTVCDSCEDSTYTQLWNWVPECLSCGSRCSSDQVETQACTREQNRICTCRPGWYCALSKQEGCRLCAPLRKCRPGFGVARPGTETSDVVCKPCAPGTFSNTTSSTDICRPHQICNVVAIPGNASMDAVCTSTSPTRSMAPGAVHLPQPVSTRSQHTQPTPEPSTAPSTSFLLPMGPSPPAEGSTGDFALPVGLIVGVTALGLLIIGVVNCVIMTQVKKKPLCLQREAKVPHLPADKARGTQGPEQQHLLITAPSSSSSSLESSASALDRRAPTRNQPQAPGVEASGAGEARASTGSSDSSPGGHGTQVNVTCIVNVCSSSDHSSQCSSQASSTMGDTDSSPSESPKDEQVPFSKEECAFRSQLETPETLLGSTEEKPLPLGVPDAGMKPS
- Purification:Protein A purified
- Cat. No.:103333-304
- Supplier no.:H00007133-D01P
The TNF RII / TNFRSF1B Antibody from Novus Biologicals is a rabbit polyclonal antibody to TNF RII / TNFRSF1B. This antibody reacts with human, mouse. The TNF RII / TNFRSF1B Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: TNF RII/TNFRSF1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse