Anti-TNF RI/TNFRSF1A Rabbit Polyclonal Antibody
103271-528
- Antibody Type:Primary
- Antigen Symbol:TNF RI/TNFRSF1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:7132
- Antigen Synonyms:p55-R|tumor necrosis factor receptor 1A isoform beta|TNF-R|tumor necrosis factor-alpha receptor|tumor necrosis factor binding protein 1|Tumor necrosis factor receptor type I|TNFR-I|TNF-R-I|tumor necrosis factor receptor type 1|TNFR55|p55|CD120a antigen|TNFR1TBP1|CD120a|FPF|tumor necrosis factor receptor superfamily|p60|TNF-R1|TNFARMGC19588|tumor necrosis factor receptor superfamily member 1A|TNF-RI|Tumor necrosis factor receptor 1|TNFR60|TNF-R55|member 1A
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:GDREKRDSVCPQGKYIHPQNNSICCTKCHKGTYLYNDCPGPGQDTDCRECESGSFTASENHLRHCLSCSKCRKEMGQVEISSCTVDRDTVCGCRKNQYRHYWSENLFQCFNCSLCLNGTVHLSCQEKQNTVCTCHAGFFLRENECVSC
- Purification:Immunogen affinity purified
- Cat. No.:103271-528
- Supplier no.:NBP1-88140
The TNF RI / TNFRSF1A Antibody from Novus Biologicals is a rabbit polyclonal antibody to TNF RI / TNFRSF1A. This antibody reacts with human. The TNF RI / TNFRSF1A Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TNF RI/TNFRSF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human