Anti-TMEM8C Rabbit Polyclonal Antibody
Catalog # 103283-290
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:TMEM8C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:389827
- Antigen Synonyms:Transmembrane Protein 8C|MYOMAKER|TMEM226|Transmembrane Protein 226
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against a recombinant protein corresponding to amino acids: KWLQKMKEKKGLYPDKSVYTQQ
- Purification:Immunogen affinity purified
- Cat. No.:103283-290
- Supplier no.:NBP2-34175
Specifications
About this item
The TMEM8C Antibody from Novus Biologicals is a rabbit polyclonal antibody to TMEM8C. This antibody reacts with human. The TMEM8C Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TMEM8C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human