To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TMEM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:7109
- Antigen Synonyms:Epilepsy holoprosencephaly candidate 1 protein|TMEM1MGC126777|Trafficking protein particle complex subunit TMEM1|TRS130|EHOC1GT334|TRAPP 130 kDa subunit|epilepsy holoprosencephaly candidate-1 protein|Protein GT334|transmembrane protein 1|TRAPP subunit TMEM1|trafficking protein particle complex subunit 10|TRS30|trafficking protein particle complex 10|trafficking protein particle complex subunit 130|EHOC-1FLJ55683|FLJ54223|Transport protein particle subunit TMEM1|FLJ54817
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TMEM1 (NP_003265.3, 1 a.a. - 1259 a.a.) full-length human protein. MDASEEPLPPVIYTMENKPIVTCAGDQNLFTSVYPTLSQQLPREPMEWRRSYGRAPKMIHLESNFVQFKEELLPKEGNKALLTFPFLHIYWTECCDTEVYKATVKDDLTKWQNVLKAHSSVDWLIVIVENDAKKKNKTNILPRTSIVDKIRNDFCNKQSDRCVVLSDPLKDSSRTQESWNAFLTKLRTLLLMSFTKNLGKFEDDMRTLREKRTEPGWSFCEYFMVQEELAFVFEMLQQFEDALVQYDELDALFSQYVVNFGAGDGANWLTFFCQPVKSWNGLILRKPIDMEKRESIQRREATLLDLRSYLFSRQCTLLLFLQRPWEVAQRALELLHNCVQELKLLEVSVPPGALDCWVFLSCLEVLQRIEGCCDRAQIDSNIAHTVGLWSYATEKLKSLGYLCGLVSEKGPNSEDLNRTVDLLAGLGAERPETANTAQSPYKKLKEALSSVEAFEKHYLDLSHATIEMYTSIGRIRSAKFVGKDLAEFYMRKKAPQKAEIYLQGALKNYLAEGWALPITHTRKQLAECQKHLGQIENYLQTSSLLASDHHLTEEERKHFCQEILDFASQPSDSPGHKIVLPMHSFAQLRDLHFDPSNAVVHVGGVLCVEITMYSQMPVPVHVEQIVVNVHFSIEKNSYRKTAEWLTKHKTSNGIINFPPETAPFPVSQNSLPALELYEMFERSPSDNSLNTTGIICRNVHMLLRRQESSSSLEMPSGVALEEGAHVLRCSHVTLEPGANQITFRTQAKEPGTYTLRQLCASVGSVWFVLPHIYPIVQYDVYSQEPQLHVEPLADSLLAGIPQRVKFTVTTGHYTIKNGDSLQLSNAEAMLILCQAESRAVVYSNTREQSSEAALRIQSSDKVTSISLPVAPAYHVIEFELEVLSLPSAPALGGESDMLGMAEPHRKHKDKQRTGRCMVTTDHKVSIDCPWSIYSTVIALTFSVPFRTTHSLLSSGTRKYVQVCVQNLSELDFQLSDSYLVDTGDSTDLQLVPLNTQSQQPIYSKQSVFFVWELKWTEEPPPSLHCRFSVGFSPASEEQLSISLKPYTYEFKVENFFTLYNVKAEIFPPSGMEYCRTGSLCSLEVLITRLSDLLEVDKDEALTESDEHFSTKLMYEVVDNSSNWAVCGKSCGVISMPVAARATHRVHMEVMPLFAGYLPLPDVRLFKYLPHHSAHSSQLDADSWIENDSLSVDKHGDDQPDSSSLKSRGSVHSACSSEHKGLPMPRLQALPAGQVFNSSSGTQVLVIPSQDDHVLEVSVT
- Purification:Protein A purified
- Cat. No.:103333-278
- Supplier no.:H00007109-D01P
Specifications
About this item
The TMEM1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TMEM1. This antibody reacts with human. The TMEM1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: TMEM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human