Anti-TLL1 Mouse Polyclonal Antibody
Catalog # 103333-198
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:TLL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:7092
- Antigen Synonyms:EC 3.4.24.19|TLLEC 3.4.24.-|ASD6|EC 3.4.24|tolloid-like protein 1|tolloid-like 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TLL1 (AAH16922.1, 1 a.a. - 392 a.a.) full-length human protein. MGLGTLSPRMLVWLVASGIVFYGELWVCAGLDYDYTFDGNEEDKTETIDYKDPCKAAVFWGDIALDDEDLNIFQIDRTIDLTQNPFGNLGHTTGGLGDHAMSKKRGALYQLIDRIRRIGFGLEQNNTVKGKVPLQFSGQNEKNRVPRAATSRTERIWPGGVIPYVIGGNFTGSQRAMFKQAMRHWEKHTCVTFIERSDEESYIVFTYRPCGCCSYVGRRGNGPQAISIGKNCDKFGIVVHELGHVIGFWHEHTRPDRDNHVTIIRENIQPGQEYNFLKMEPGEVNSLGERYDFDSIMHYARNTFSRGMFLDTILPSRDDNGIRPAIGQRTRLSKGDIAQARKLYRCPACGETLQESNGNLSSPGFPNGYPSYTHCIWRVSVTPGEKVVFSLC
- Purification:Protein G purified
- Cat. No.:103333-198
- Supplier no.:H00007092-B01P
Specifications
About this item
The TLL1 Antibody from Novus Biologicals is a mouse polyclonal antibody to TLL1. This antibody reacts with human. The TLL1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: TLL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human