Anti-TFB1M Mouse Polyclonal Antibody
Catalog # 103343-674
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:TFB1M
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:51106
- Antigen Synonyms:Mitochondrial 12S rRNA dimethylase 1|homolog of yeast mitochondrial transcription factor B|N'-adenosyl(rRNA) dimethyltransferase 1|dimethyladenosine transferase 1|S-adenosylmethionine-6-N'|mtTFB|CGI-75|EC 2.1.1.-|h-mtTFB1|mitochondrial|CGI75|EC 2.1.1|Mitochondrial transcription factor B1|transcription factor B1|h-mtTFB|hTFB1M|mtTFB1|mitochondrial dimethyladenosine transferase 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TFB1M (AAH17788.1, 1 a.a. ~ 346 a.a) full-length human protein.MAASGKLSTCRLPPLPTIREIIKLLRLQAAKQLSQNFLLDLRLTDKIVRKAGNLTNAYVYEVGPGPGGITRSILNADVAELLVVEKDTRFIPGLQMLSDAAPGKLRIVHGDVLTFKVEKAFSESLKRPWEDDPPNVHIIGNLPFSVSTPLIIKWLENISCRDGPFVYGRTQMTLTFQKEVAERLAANTGSKQRSRLSVMAQYLCNVRHIFTIPGQAFVPKPEVDVGVVHFTPLIQPKIEQPFKLVEKVVQNVFQF
- Purification:Protein A purified
- Cat. No.:103343-674
- Supplier no.:H00051106-B02P
Specifications
About this item
The TFB1M Antibody from Novus Biologicals is a mouse polyclonal antibody to TFB1M. This antibody reacts with human. The TFB1M Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: TFB1M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human