Anti-TFAP2D Goat Polyclonal Antibody
Catalog # 103409-246
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:TFAP2D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 μg
- Epitope:internal region
- Gene ID:83741
- Antigen Synonyms:AP2-delta|TFAP2BL1activating enhancer binding protein 2 beta-like 1|Transcription factor AP-2-beta-like 1|Activating enhancer-binding protein 2-delta|AP-2 like|transcription factor AP-2 beta (activating enhancer binding protein 2beta)-like 1|transcription factor AP-2 beta (activating enhancer-binding protein 2beta)-like 1|transcription factor AP-2-delta|transcription factor AP-2 delta (activating enhancer binding protein 2 delta)
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-HHQSFHYEFQHSH corresponding to internal region according to NP_758438.2.
- Tested Applications:Peptide
- Purification:Immunogen affinity purified
- Cat. No.:103409-246
Specifications
About this item
The TFAP2D Antibody from Novus Biologicals is a goat polyclonal antibody to TFAP2D. This antibody reacts with human, mouse. The TFAP2D Antibody has been validated for the following applications: Peptide ELISA.
Type: Primary
Antigen: TFAP2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal region
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse