Anti-TESK2 Mouse Monoclonal Antibody [clone: 5H4]
Catalog # 103338-858
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:TESK2
- Clonality:Monoclonal
- Clone:5H4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:10420
- Antigen Synonyms:testis-specific kinase 2|Testicular protein kinase 2|dual specificity testis-specific protein kinase 2|EC 2.7.12.1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TESK2 (AAH33085, 405 a.a. ~ 542 a.a) partial recombinant protein with GST tag.GPGTMPLADWQEPLAPPIRRWCSLPGSPEFLHQEACPFVGREESLSDGPPPRLSSLKYRVKEIPPFRASALPAAQAHEAMDCSILQEENGFGSRPQGTSPCPAGASEEMEVEERPAGSTPATFSTSGIGLQTQGKQDG
- Purification:IgG purified
- Cat. No.:103338-858
- Supplier no.:H00010420-M04
Specifications
About this item
The TESK2 Antibody (5H4) from Novus Biologicals is a mouse monoclonal antibody to TESK2. This antibody reacts with human. The TESK2 Antibody (5H4) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TESK2
Clonality: Monoclonal
Clone: 5H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human