Anti-TAF9b Mouse Polyclonal Antibody
Catalog # 103344-338
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:TAF9b
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:51616
- Antigen Synonyms:TAF9B RNA polymerase II|transcription initiation factor TFIID subunit 9B|Neuronal cell death-related protein 7|TFIID-31|TATA box binding protein (TBP)-associated factor|DN-7Transcription-associated factor TAFII31L|31kDa|TAF9Ltranscription associated factor TAFII31L|transcription initiation factor IID|31kD subunit|DN7|TAF9-like RNA polymerase II|TAFII31LTBP-associated factor 9L|Transcription initiation factor TFIID subunit 9-like
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TAF9B (NP_057059.2, 1 a.a. ~ 251 a.a) full-length human protein.
- Purification:Protein G purified
- Cat. No.:103344-338
- Supplier no.:H00051616-B01P
Specifications
About this item
The TAF9b Antibody from Novus Biologicals is a mouse polyclonal antibody to TAF9b. This antibody reacts with human. The TAF9b Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: TAF9b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human