To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:SUSD2
- Clonality:Monoclonal
- Clone:1279B
- Conjugation:DyLight® 488
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:56241
- Antigen Synonyms:sushi domain-containing protein 2|BK65A6.2|FLJ22778|Sushi domain (SCR repeat) containing|sushi domain containing 2
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Partial recombinant human SUSD2 protein (amino acids 1-785) [UniProt Q9UGT4]
- Purification:Protein A or G purified
- Cat. No.:103294-254
- Supplier no.:MAB90564G
Specifications
About this item
The SUSD2 Antibody (1279B) [DyLight 488] from Novus Biologicals is a rabbit monoclonal antibody to SUSD2. This antibody reacts with human, rat. The SUSD2 Antibody (1279B) [DyLight 488] has been validated for the following applications: Western Blot, Flow Cytometry.
Type: Primary
Antigen: SUSD2
Clonality: Monoclonal
Clone: 1279B
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat