Anti-SUSD2 Rabbit Monoclonal Antibody [clone: 1279B]
103292-618
- Antibody Type:Primary
- Antigen Symbol:SUSD2
- Clonality:Monoclonal
- Clone:1279B
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 ug
- Gene ID:56241
- Antigen Synonyms:sushi domain-containing protein 2|BK65A6.2|FLJ22778|Sushi domain (SCR repeat) containing|sushi domain containing 2
- Storage Buffer:PBS
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Partial recombinant human SUSD2 protein (amino acids 1-785) [UniProt Q9UGT4]
- Tested Applications:, CyTOF-ready
- Purification:Protein A or G purified
- Cat. No.:103292-618
- Supplier no.:MAB90564
The SUSD2 Antibody (1279B) - Azide Free from Novus Biologicals is a rabbit monoclonal antibody to SUSD2. This antibody reacts with human, rat. The SUSD2 Antibody (1279B) - Azide Free has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry (Negative), ICC / IF (Negative), CyTOF-ready.
Type: Primary
Antigen: SUSD2
Clonality: Monoclonal
Clone: 1279B
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat