Order Entry
United States
ContactUsLinkComponent
Anti-SPSB3 Mouse Polyclonal Antibody
Anti-SPSB3 Mouse Polyclonal Antibody
  103345-748
Anti-SPSB3 Mouse Polyclonal Antibody
  103345-748
 :  H00090864-B01P

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    SPSB3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.05 mg
  • Gene ID:
    90864
  • Antigen Synonyms:
    SSB3SPRY domain-containing SOCS box protein 3|SSB-3|chromosome 16 open reading frame 31|splA/ryanodine receptor domain and SOCS box containing 3|SPRY domain-containing SOCS box protein SSB-3|C16orf31
  • Storage Buffer:
    PBS (pH 7.4)
  • Storage Temperature:
    Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
  • Immunogen:
    SPSB3 (AAH24244.1, 1 a.a. ~ 355 a.a) full-length human protein.MARRPRNSRAWHFVLSAARRDADARAVALAGSTNWGYDSDGQHSDSDSDPEYSTLPPSIPSAVPVTGESFCDCAGQSEASFCSSLHSAHRGRDCRCGEEDEYFDWVWDDLNKSSATLLSCDNRKVSFHMEYSCGTAAIRGTKELGEGQHFWEIKMTSPVYGTDMMVGIGTSDVDLDKYRHTFCSLLGRDEDSWGLSYTGLLHHKGDKTSFSSRFGQGSIIGVHLDTWHGTLTFFKNRKCIGVAATKLQNKRFYPM
  • Purification:
    Protein A purified
  • Cat. No.:
    103345-748
  • Supplier no.:
    H00090864-B01P

 

 

The SPSB3 Antibody from Novus Biologicals is a mouse polyclonal antibody to SPSB3. This antibody reacts with human. The SPSB3 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: SPSB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human