Anti-SOX8 Mouse Monoclonal Antibody [clone: 8G7]
Catalog # 103343-302
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:SOX8
- Clonality:Monoclonal
- Clone:8G7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:30812
- Antigen Synonyms:transcription factor SOX-8|SRY (sex determining region Y)-box 8|MGC24837
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SOX8 (NP_055402, 102 a.a. ~ 211 a.a) partial recombinant protein with GST tag.VKRPMNAFMVWAQAARRKLADQYPHLHNAELSKTLGKLWRLL SESEKRPFVEEAERLRVQHKKDHPDYKYQPRRRKSAKAGHSD SDSGAELGPHPGGGAVYKAEAGLGD
- Purification:IgG purified
- Cat. No.:103343-302
- Supplier no.:H00030812-M01
Specifications
About this item
The SOX8 Antibody (8G7) from Novus Biologicals is a mouse monoclonal antibody to SOX8. This antibody reacts with human. The SOX8 Antibody (8G7) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: SOX8
Clonality: Monoclonal
Clone: 8G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human